Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047907.3 | NP_001041372.1 | 1 - 2 | 98 | 62 | |
GenBank mRNA | DQ480748.1 | - | 1 - 2 | 102 | 62 | |
FM031503.1 | - | 1 - 2 | 98 | 62 | ||
FQ229123.1 | - | 1 - 2 | 116 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |