Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam84a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127299.1 | NP_001120771.1 | 3 - 4 | 570 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |