Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Isg20l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007741.1 | NP_001007742.1 | 2 - 3 | 1188 | 81 | |
GenBank mRNA | BC079314.1 | - | 2 - 3 | 1188 | 7134903 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |