Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099494.1 | NP_001092964.1 | 1 - 2 | 290 | 132 | |
XM_006241129.1 | XP_006241191.1 | 2 - 3 | 509 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |