Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009491.1 | NP_001009491.1 | 4 - 5 | 928 | 80 | |
XM_008769247.1 | XP_008767469.1 | 4 - 5 | 1005 | 80 | ||
GenBank mRNA | AY196701.1 | - | 4 - 5 | 928 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |