Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pomgnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766681.1 | XP_008764903.1 | 2 | 2372 | 139 | |
XM_008766682.1 | XP_008764904.1 | 2 | 3737 | 139 | ||
XM_008766683.1 | XP_008764905.1 | 3 | 2442 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |