Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013052.1 | NP_001013070.1 | 1 | 666 | 95 | |
XM_006253021.2 | XP_006253083.1 | 2 | 2863 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon |