Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aste1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040156.2 | NP_001035246.2 | 4 - 5 | 1967 | 153 | |
XM_006243672.2 | XP_006243734.1 | 3 - 4 | 1722 | 153 | ||
XM_006243675.2 | XP_006243737.1 | 4 - 5 | 1614 | 153 | ||
XM_008766543.1 | XP_008764765.1 | 3 - 4 | 2151 | 153 | ||
XM_008766544.1 | XP_008764766.1 | 3 - 4 | 2151 | 153 | ||
XM_008766545.1 | XP_008764767.1 | 3 - 4 | 846 | 153 | ||
XM_008766546.1 | XP_008764768.1 | 3 - 4 | 492 | 153 | ||
XM_008766547.1 | XP_008764769.1 | 2 - 3 | 342 | 153 | ||
XM_008766548.1 | XP_008764770.1 | 3 - 4 | 386 | 153 | ||
GenBank mRNA | BC083913.1 | - | 4 - 5 | 514 | 153 | |
BQ193960.1 | - | 1 - 2 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Probe spans exons |