Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY325173.1 | - | 495 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus |