Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom2r68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099481.1 | NP_001092951.1 | 4 - 5 | 1523 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Probe spans exons |