Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100534.1 | NP_001094004.1 | 4 - 5 | 782 | 109 | |
GenBank mRNA | BC159423.1 | - | 4 - 5 | 670 | 109 | |
BC159431.1 | - | 4 - 5 | 782 | 9091211 | 109 | |
FQ211668.1 | - | 3 - 4 | 477 | 109 | ||
FQ217699.1 | - | 3 - 4 | 477 | 109 | ||
FQ219786.1 | - | 3 - 4 | 477 | 109 | ||
FQ220709.1 | - | 3 - 4 | 477 | 109 | ||
FQ220767.1 | - | 3 - 4 | 477 | 109 | ||
FQ221206.1 | - | 3 - 4 | 476 | 109 | ||
FQ221432.1 | - | 3 - 4 | 477 | 109 | ||
FQ221889.1 | - | 3 - 4 | 109 | |||
FQ222645.1 | - | 3 - 4 | 477 | 109 | ||
FQ222683.1 | - | 3 - 4 | 477 | 109 | ||
FQ223813.1 | - | 3 - 4 | 477 | 109 | ||
FQ225844.1 | - | 3 - 4 | 477 | 109 | ||
FQ226679.1 | - | 3 - 4 | 477 | 109 | ||
FQ228074.1 | - | 3 - 4 | 477 | 109 | ||
FQ228176.1 | - | 3 - 4 | 477 | 109 | ||
FQ229585.1 | - | 3 - 4 | 477 | 109 | ||
FQ229680.1 | - | 3 - 4 | 477 | 109 | ||
FQ229824.1 | - | 3 - 4 | 477 | 109 | ||
FQ230037.1 | - | 3 - 4 | 477 | 109 | ||
FQ231936.1 | - | 3 - 4 | 477 | 109 | ||
FQ232074.1 | - | 3 - 4 | 477 | 109 | ||
FQ232741.1 | - | 3 - 4 | 477 | 109 | ||
FQ235314.1 | - | 3 - 4 | 477 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |