Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108598.2 | NP_001102068.2 | 5 - 6 | 466 | 111 | |
GenBank mRNA | CB576376.1 | - | 5 - 6 | 343 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Detect Genomic DNA Rattus norvegicus Probe spans exons |