Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212516.2 | NP_997681.1 | 2 - 3 | 241 | 76 | |
GenBank mRNA | BC058497.1 | - | 2 - 3 | 242 | 6918545 | 76 |
FQ210248.1 | - | 2 - 3 | 231 | 76 | ||
FQ211867.1 | - | 2 - 3 | 238 | 76 | ||
FQ214576.1 | - | 237 | 76 | |||
FQ217139.1 | - | 2 - 3 | 257 | 76 | ||
FQ217410.1 | - | 2 - 3 | 258 | 76 | ||
FQ217906.1 | - | 2 - 3 | 230 | 76 | ||
FQ218008.1 | - | 2 - 3 | 230 | 76 | ||
FQ223878.1 | - | 2 - 3 | 231 | 76 | ||
FQ223920.1 | - | 2 - 3 | 237 | 76 | ||
FQ224004.1 | - | 2 - 3 | 231 | 76 | ||
FQ224067.1 | - | 2 - 3 | 231 | 76 | ||
FQ224620.1 | - | 2 - 3 | 231 | 76 | ||
FQ229120.1 | - | 2 - 3 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |