Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001184574.1 | NP_878154.1 | 1 | 74 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Saccharomyces cerevisiae Both primers and probe map within a single exon |