Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAGLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244150.1 | NP_001231079.1 | 3 | 372 | 73 | |
XM_005667370.3 | XP_005667427.1 | 498 | 73 | |||
GenBank mRNA | AF053629.1 | - | 2 | 152 | 73 | |
AK231706.1 | - | 3 | 285 | 73 | ||
AK231867.1 | - | 3 | 281 | 73 | ||
AK234135.1 | - | 3 | 336 | 73 | ||
AK236759.1 | - | 3 | 281 | 73 | ||
AK238084.1 | - | 3 | 286 | 73 | ||
AK345464.1 | - | 3 | 256 | 73 | ||
AK393811.1 | - | 3 | 286 | 73 | ||
AK393849.1 | - | 3 | 285 | 73 | ||
AK397718.1 | - | 3 | 411 | 73 | ||
AK397931.1 | - | 3 | 286 | 73 | ||
AK398562.1 | - | 3 | 285 | 73 | ||
AK399019.1 | - | 3 | 308 | 73 | ||
AK400885.1 | - | 3 | 285 | 73 | ||
AY610393.1 | - | 3 | 259 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Inventoried Amplicon spans exons and probe does not span exons |