Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003359865.4 | XP_003359913.1 | 1127 | 82 | ||
XM_005672653.2 | XP_005672710.1 | 1218 | 82 | |||
XM_005672654.2 | XP_005672711.1 | 1057 | 82 | |||
GenBank mRNA | AM410991.1 | - | 1 | 150 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Both primers and probe map within a single exon |