Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112689.1 | NP_001106159.2 | 2 - 3 | 96 | 78 | |
XM_005669744.3 | XP_005669801.1 | 231 | 78 | |||
XM_005669745.3 | XP_005669802.1 | 215 | 78 | |||
XM_013981921.1 | XP_013837375.1 | 129 | 78 | |||
XM_013981922.2 | XP_013837376.1 | 119 | 78 | |||
GenBank mRNA | AK232875.1 | - | 2 - 3 | 108 | 78 | |
AK390817.1 | - | 2 - 3 | 232 | 78 | ||
EF374062.1 | - | 2 - 3 | 96 | 78 | ||
EU395789.1 | - | 1 - 2 | 60 | 78 | ||
JF923469.1 | - | 1 - 2 | 50 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |