Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213885.1 | NP_999050.1 | 8 - 9 | 1650 | 103 | |
XM_021082358.1 | XP_020938017.1 | 1887 | 103 | |||
GenBank mRNA | Y12503.1 | - | 1650 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |