Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001633.2 | NP_001001633.2 | 2 - 3 | 225 | 71 | |
GenBank mRNA | AK232797.1 | - | 2 - 3 | 196 | 71 | |
AK233927.1 | - | 2 - 3 | 196 | 71 | ||
AK235789.1 | - | 2 - 3 | 197 | 71 | ||
AK344834.1 | - | 2 - 3 | 198 | 71 | ||
AK347458.1 | - | 2 - 3 | 201 | 71 | ||
AK348176.1 | - | 2 - 3 | 201 | 71 | ||
AK399710.1 | - | 198 | 71 | |||
AK399903.1 | - | 2 - 3 | 226 | 71 | ||
AY550074.1 | - | 2 - 3 | 166 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |