Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033013.2 | NP_001028185.1 | 1 - 2 | 209 | 87 | |
XM_005663157.3 | XP_005663214.1 | 220 | 87 | |||
XM_013996818.2 | XP_013852272.1 | 221 | 87 | |||
XM_013996819.2 | XP_013852273.1 | 220 | 87 | |||
GenBank mRNA | AK233490.1 | - | 1 - 2 | 209 | 87 | |
AK239470.1 | - | 1 - 2 | 208 | 87 | ||
AK391270.1 | - | 1 - 2 | 214 | 87 | ||
DQ026064.1 | - | 1 - 2 | 212 | 87 | ||
DQ027013.1 | - | 1 - 2 | 212 | 87 | ||
FJ608551.1 | - | 1 - 2 | 114 | 87 | ||
HM122239.1 | - | 1 - 2 | 140 | 87 | ||
HM803104.1 | - | 1 - 2 | 140 | 87 | ||
HM803105.1 | - | 1 - 2 | 140 | 87 | ||
KF595284.1 | - | 1 - 2 | 116 | 87 |
Gene Symbol: LOC110260307 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021089520.1 | XP_020945179.1 | 223 | 87 | ||
XM_021089523.1 | XP_020945182.1 | 222 | 87 | |||
XM_021089524.1 | XP_020945183.1 | 222 | 87 | |||
GenBank mRNA | AK347675.1 | - | 1 - 2 | 214 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |