Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYROBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214202.1 | NP_999367.1 | 4 - 5 | 272 | 89 | |
GenBank mRNA | AF152021.1 | - | 4 - 5 | 272 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |