Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214216.1 | NP_999381.1 | 2 - 3 | 192 | 96 | |
XM_021088713.1 | XP_020944372.1 | 307 | 96 | |||
XM_021088714.1 | XP_020944373.1 | 359 | 96 | |||
XM_021088715.1 | XP_020944374.1 | 3337 | 96 | |||
GenBank mRNA | AF056489.1 | - | 2 - 3 | 192 | 96 | |
AK344681.1 | - | 3 - 4 | 306 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |