Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166044.1 | NP_001159516.1 | 3 - 4 | 263 | 66 | |
GenBank mRNA | AK235860.1 | - | 3 - 4 | 384 | 66 | |
GQ449385.1 | - | 3 - 4 | 263 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |