Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166044.1 | NP_001159516.1 | 5 - 6 | 608 | 82 | |
GenBank mRNA | AK235860.1 | - | 5 - 6 | 729 | 82 | |
GQ449385.1 | - | 5 - 6 | 608 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Sus scrofa Probe spans exons |