Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190251.1 | NP_001177180.1 | 5 - 6 | 548 | 68 | |
GenBank mRNA | AB490118.1 | - | 4 - 5 | 508 | 68 | |
AY610013.1 | - | 5 - 6 | 566 | 68 | ||
FJ469911.1 | - | 4 - 5 | 451 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |