Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195325.1 | NP_001182254.1 | 3 - 4 | 599 | 65 | |
GenBank mRNA | GU373709.1 | - | 3 - 4 | 599 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Sus scrofa Probe spans exons |