Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204382.1 | NP_001191311.1 | 4 - 5 | 404 | 57 | |
XM_021090906.1 | XP_020946565.1 | 638 | 57 | |||
XM_021090908.1 | XP_020946567.1 | 645 | 57 | |||
XM_021090910.1 | XP_020946569.1 | 368 | 57 | |||
XM_021090911.1 | XP_020946570.1 | 583 | 57 | |||
XM_021090913.1 | XP_020946572.1 | 368 | 57 | |||
GenBank mRNA | AK239479.1 | - | 5 - 6 | 351 | 57 | |
AK350790.1 | - | 4 - 5 | 404 | 57 | ||
AK391116.1 | - | 4 - 5 | 560 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Sus scrofa Probe spans exons |