Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243352.1 | NP_001230281.1 | 3 - 4 | 619 | 117 | |
XM_005654033.3 | XP_005654090.1 | 3309 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |