Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246205.1 | NP_001233134.1 | 6 - 7 | 1105 | 123 | |
GenBank mRNA | AK233687.1 | - | 7 - 8 | 1243 | 123 | |
AK391273.1 | - | 7 - 8 | 1210 | 123 | ||
HQ266637.1 | - | 6 - 7 | 1105 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Sus scrofa Probe spans exons |