Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206447.1 | NP_001193376.1 | 4 - 5 | 1598 | 96 | |
GenBank mRNA | AK351123.1 | - | 4 - 5 | 1596 | 96 | |
AK389431.1 | - | 4 - 5 | 1025 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |