Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206456.1 | NP_001193385.1 | 4 - 5 | 827 | 71 | |
GenBank mRNA | CV874961.1 | - | 2 - 3 | 324 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |