Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243397.1 | NP_001230326.1 | 1 - 2 | 277 | 95 | |
XM_005652508.3 | XP_005652565.1 | 346 | 95 | |||
GenBank mRNA | AK344907.1 | - | 1 - 2 | 280 | 95 | |
BP441548.1 | - | 1 - 2 | 277 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |