Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243478.1 | NP_001230407.1 | 5 - 6 | 720 | 80 | |
GenBank mRNA | AK231488.1 | - | 5 - 6 | 722 | 80 | |
AK240399.1 | - | 5 - 6 | 639 | 80 | ||
AK346690.1 | - | 5 - 6 | 639 | 80 | ||
AY610201.1 | - | 5 - 6 | 696 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |