Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243701.1 | NP_001230630.1 | 6 - 7 | 1821 | 62 | |
GenBank mRNA | AK346293.1 | - | 6 - 7 | 1821 | 62 | |
AK390410.1 | - | 6 - 7 | 1948 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |