Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243711.1 | NP_001230640.1 | 3 - 4 | 439 | 75 | |
XM_005660986.2 | XP_005661043.1 | 736 | 75 | |||
XM_005660987.3 | XP_005661044.1 | 579 | 75 | |||
XM_021077723.1 | XP_020933382.1 | 428 | 75 | |||
GenBank mRNA | AK349798.1 | - | 3 - 4 | 744 | 75 | |
AK396456.1 | - | 3 - 4 | 433 | 75 | ||
AK400493.1 | - | 3 - 4 | 527 | 75 | ||
FS675912.1 | - | 3 - 4 | 451 | 75 | ||
FS721678.1 | - | 3 - 4 | 439 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |