Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244502.1 | NP_001231431.1 | 5 - 6 | 405 | 63 | |
GenBank mRNA | EV860936.2 | - | 1 - 2 | 65 | 63 | |
FD593970.1 | - | 5 - 6 | 407 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Sus scrofa Probe spans exons |