Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY351862.1 | - | 8 - 9 | 722 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |