Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VVME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | U67426.1 | - | 2 - 3 | 459 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Vitis vinifera Probe spans exons |