Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | GQ161924.1 | - | 7 - 8 | 762 | 92 | |
GQ161925.1 | - | 7 - 8 | 762 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Vitis vinifera Probe spans exons |