Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsto2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005086.2 | NP_001005086.1 | 2 - 3 | 212 | 95 | |
XM_012966444.2 | XP_012821898.1 | 3 - 4 | 286 | 95 | ||
XM_012966445.1 | XP_012821899.1 | 2 - 3 | 271 | 95 | ||
GenBank mRNA | BC077010.1 | - | 2 - 3 | 213 | 95 | |
CR760625.2 | - | 2 - 3 | 186 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |