Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203514.1 | NP_988845.1 | 4 - 5 | 362 | 65 | |
GenBank mRNA | BC063212.1 | - | 4 - 5 | 362 | 65 | |
BC075541.1 | - | 4 - 5 | 426 | 65 | ||
CR762283.2 | - | 4 - 5 | 316 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |