Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203823.1 | NP_989154.1 | 2 - 3 | 378 | 84 | |
GenBank mRNA | BC061629.1 | - | 2 - 3 | 378 | 84 | |
BC080971.1 | - | 2 - 3 | 378 | 84 | ||
CR942746.2 | - | 2 - 3 | 373 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |