Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lamtor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203972.1 | NP_989303.1 | 2 - 3 | 372 | 67 | |
GenBank mRNA | BC064269.1 | - | 2 - 3 | 372 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |