Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005120.2 | NP_001005120.2 | 1 - 2 | 196 | 77 | |
GenBank mRNA | BC077060.1 | - | 2 - 3 | 77 | ||
CR573361.1 | - | 1 - 2 | 196 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |