Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nipsnap3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005718.1 | NP_001005718.1 | 5 - 6 | 808 | 66 | |
GenBank mRNA | BC075307.1 | - | 5 - 6 | 808 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |