Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rexo4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006867.1 | NP_001006868.1 | 5 - 6 | 1116 | 82 | |
GenBank mRNA | BC076977.1 | - | 5 - 6 | 1116 | 82 | |
CR855796.2 | - | 5 - 6 | 1094 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |