Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rraga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006896.1 | NP_001006897.1 | 4 - 5 | 262 | 73 | |
GenBank mRNA | BC074593.1 | - | 4 - 5 | 262 | 73 | |
CR926416.2 | - | 3 - 4 | 225 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |