Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: degs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007484.1 | NP_001007485.1 | 2 - 3 | 868 | 103 | |
XM_012962906.2 | XP_012818360.1 | 2 - 3 | 972 | 103 | ||
GenBank mRNA | BC079924.1 | - | 2 - 3 | 868 | 103 | |
CR855730.2 | - | 2 - 3 | 860 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |