Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cbr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007872.1 | NP_001007873.1 | 2 - 3 | 176 | 97 | |
XM_012960249.2 | XP_012815703.1 | 2 - 3 | 254 | 97 | ||
XM_012960250.2 | XP_012815704.1 | 2 - 3 | 403 | 97 | ||
XM_012960257.2 | XP_012815711.1 | 2 - 3 | 339 | 97 | ||
GenBank mRNA | BC080139.1 | - | 2 - 3 | 176 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |