Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011063.1 | NP_001011063.1 | 3 - 4 | 294 | 98 | |
GenBank mRNA | BC084442.1 | - | 3 - 4 | 294 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |